Learn More
Description
- Purity: >98% by SDS-PAGE and HPLC analysis
- Species: Rat
- Molecular weight: 18.7kd
- Contains 160 amino acid residues
- Lyophilized from 5mM Tris, pH: 7.2
- ED(50) was determined by dose-dependent inhibition of antigen-specific T-cell proliferation is <10ng/mL
Recommended Storage
Store at -20°C
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 18.7kd |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C |
| Product Type | Rat Interleukin-10 Recombinant Protein |
| Species | Rat |
| Recombinant | Recombinant |
| Purity or Quality Grade | >98% by SDS-PAGE and HPLC Analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.