Learn More
Description
- Lyophilized powder
- Biological activity: Concentration range of 0.1 to 10.0ng/mL
- Purity: >98% by SDS-PAGE and HPLC analysis
- Species: Rat
- Molecular weight: 14kd
- Contains 121 amino acid residues
Recommended Storage
Store at -20°C
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 14kd |
| Quantity | 20 μg |
| Storage Requirements | Store at -20°C |
| Product Type | Rat Interleukin-4 Recombinant Protein |
| Species | Rat |
| Recombinant | Recombinant |
| Purity or Quality Grade | >98% by SDS-PAGE and HPLC Analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.