Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Recombinant Human Pigment Epithelium Derived Factor Protein

Catalog No. GF134 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF134 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF134 Supplier MilliporeSigma™ Supplier No. GF134
Add to Cart
Add to Cart

a 50 kDa secreted protein in conditioned medium from cultured fetal human retinal pigment epithelium (RPE) cells

Pigment epithelium-derived factor (PEDF) was first identified as a 50 kDa secreted protein in conditioned medium from cultured fetal human retinal pigment epithelium (RPE) cells (Tombran-Tink et al., 1991). PEDF has been shown to have neurotrophic activity in the retina in vivo (Cayouette et al., 1999). PEDF is also a neurotrophic and neuroprotective factor in other systems, including cultured cerebellar granule cells, primary hippocampal neurons, spinal cord motor neurons and amphibian photoreceptors cultured in the absence of RPE cells (Karakousis, et al., 2001). The binding of PEDF to retinoblastoma and cerebellar granule cells may be receptor-mediated. Recently, PEDF was shown to be a potent inhibitor of angiogenesis in endothelial cell cultures and in rat cornea and retina in vivo (Dawson et al., 2001; Stellmach et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 10 μg
Source Stable BHK (baby hamster kidney) cell transfectants
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.