Learn More
MilliporeSigma™ Chemicon™ Rho-associated Kinase (ROCK) Activity Assay Kit
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Description | Rho-associated Kinase (ROCK) Assay kit |
| For Use With (Application) | For detection of the active ROCK and DMPK family kinases |
| Includes | Active Rho-associated Kinase II, MYPT1 Pre-coated 96-well multistrip plate, Anti-phospho-MYPT1(Thr696) antibody, TMB/E substrate, Stop solution; TBS, 20X, 20% Tween™ 20 (v/v); Plate sealers, Assay dilution buffer I, ATP (10mM), MgCl2 (1M), 30% BSA, Goat anti-rabbit IgG HRP Secondary antibody, ROCK inhibitor Y-27632 |
| Quantity | 96 Tests |