Learn More
MilliporeSigma™ Chemicon™ uPA Colorimetric Substrate, ECM600, ECM610
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Characterization | >95% by HPLC |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | 498.9 |
| Name | uPA Colorimetric Substrate, ECM600, ECM610 |
| Quantity | 5 mg |
| Storage Requirements | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.