Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cholesterol Esterase, Microbial, Protein

Catalog No. 2281801000U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-818-01000U 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-818-01000U Supplier MilliporeSigma™ Supplier No. 2281801000U
Add to Cart
Add to Cart
Native, microbial cholesterol esterase. Catalyzes hydrolysis and formation of cholesterol esters, ranging from cholesterol acetate to cholesterol stearate.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method 10mM potassium phosphate/sodium hydroxide, 0.1% BSA, pH 7.5.
Quantity 1000 U
Immunogen Cholesterol Esterase
Storage Requirements Reconstitute just prior to use. Enzyme will lose activity in solution upon storage.
Regulatory Status RUO
Recombinant Native
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.