Learn More
MilliporeSigma™ Chromolith™ Phenyl Guard Cartridge
Catalog No. M1520590001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
M1520590001 3/Pk.
1 options
Catalog No. M1520590001 Supplier MilliporeSigma™ Supplier No. 1520590001
Add to Cart
Add to Cart
Added directly in front of the main column to protect it from chemical or mechanical contamination
  • For use with classical particulate columns
  • Used as trap columns when large sample volumes are to be injected
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3/Pk.
Length (Metric) 5 mm
Type HPLC Guard Cartridge
Diameter (Metric) 4.6 mm
Sorbent Phenyl