Learn More
MilliporeSigma™ Chromolith™ Prep HPLC Guard Cartridges
Catalog No. M1252600001 Shop All MilliporeSigma Products
Change view
Click to view available options
Prep RP-18 endcapped sorbent
Catalog No.
M1252600001 Prep RP-18 endcapped sorbent
1 options
Catalog No. M1252600001 Supplier MilliporeSigma™ Supplier No. 1.25260.0001
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For prolonging the lifespan of Chromolith™ analytical columns
Although the Chromolith™ columns are well known for their robustness and longevity, guard columns are available to prolong the lifespan of Chromolith™ analytical columns even further. These 10-25mm guard cartridges require a holder - see part number 125262.0001
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 10 mm
Type HPLC Guard Cartridge
Diameter (Metric) Inner 25 mm
Sorbent Prep RP-18 endcapped sorbent