Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chromolith™ RP Endcapped HPLC Guard Cartridges

Catalog No. p-4752666 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric) Inner:
2 mm
3 mm
4.6 mm
Length (Metric):
5 mm
10 mm
Sorbent:
RP-18 endcapped sorbent
RP-8 endcapped sorbent
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Length (Metric) Sorbent
M1514520001 4.6 mm 10 mm RP-18 endcapped sorbent
M1514500001 4.6 mm 5 mm RP-18 endcapped sorbent
M1520050001 3 mm 5 mm RP-18 endcapped sorbent
M1520130001 4.6 mm 5 mm RP-8 endcapped sorbent
M1520090001 2 mm 5 mm RP-18 endcapped sorbent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1514520001 Supplier MilliporeSigma™ Supplier No. 1.51452.0001
Only null left
Add to Cart
Add to Cart

Also popular for the protection of classical packed HPLC columns

  • Handles easily
  • Adds to the robustness and longevity of monolithic analytical columns

TRUSTED_SUSTAINABILITY

Specifications

Quantity 3/Pk.
Length (Metric) 10 mm
Type HPLC Guard Cartridge
Diameter (Metric) Inner 4.6 mm
Sorbent RP-18 endcapped sorbent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.