Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Coagulation Factor IX, Human Plasma, Protein

Catalog No. 233279100U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-327-9100U 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-327-9100U Supplier MilliporeSigma™ Supplier No. 233279100U
Add to Cart
Add to Cart
Native coagulation factor IX from human plasma. Sold on the basis of plasma equivalent units. 100 plasma equivalent units are equal to 500μg of Factor IX. In vivo, activation of Factor IX to IXa occurs via either the tissue factor/Factor VIIa complex (the extrinsic pathway), or Factor XIa (the intrinsic pathway). Activated Factor IXa, in a complex with Factor VIIIa, can then convert Factor X to Xa, the catalytic component of the prothrombinase complex.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 20mM Tris-HCl, 1mM benzamidine, pH 7.5.
Purification Method SDS-PAGE
Quantity 100 U
Immunogen Coagulation Factor IX
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.