Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Coagulation Factor VIIa, Human Plasma, Protein

Catalog No. 21937010UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-937-010UG 10μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-937-010UG Supplier MilliporeSigma™ Supplier No. 21937010UG
Add to Cart
Add to Cart
Native coagulation factor VIIa from human plasma prepared from human Factor VII using human Factor XIIa, which was removed after activation. In the coagulation process, after complexing with tissue factor and membrane phospholipids, Factor VIIa activates Factors IX and X to the enzymatically active Factors IXa and Xa. Provides a crossover point between the extrinsic and intrinsic pathways.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 20mM Tris-HCl, pH 7.5.
Purification Method SDS-PAGE
Quantity 10μg
Immunogen Coagulation Factor VIIa
Storage Requirements Following initial use aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.