Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 700mM NaCl, 20mM Tris-HCl, pH 7.4. |
| Preparation Method | H2O. |
| Purification Method | Homogeneous by SDS-PAGE |
| Quantity | 10 U |
| Immunogen | Coagulation Factor Xa |
| Storage Requirements | Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.