Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Coagulation Factor XII, Human Plasma, Protein

Catalog No. 233490500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-349-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-349-0500UG Supplier MilliporeSigma™ Supplier No. 233490500UG
Add to Cart
Add to Cart
Native coagulation factor XII from human plasma. A single-chain polypeptide which, once activated (principally from the action of kallikrein), is converted to Factor α-XIIa. Factor α-XIIa then converts Factor XI to Factor XIa.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 4mM sodium acetate buffer, pH 5.3.
Purification Method Single band by SDS-PAGE
Quantity 500 μg
Immunogen Coagulation Factor XII
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.