Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cobra Venom Factor, Naja naja kaouthia, Protein

Catalog No. 2335521MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-355-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-355-21MG Supplier MilliporeSigma™ Supplier No. 2335521MG
Add to Cart
Add to Cart
Native cobra venom factor from Naja naja kaouthia. Glycoprotein composed of three non-identical disulfide-bonded subunits with M.W. of 68kDa (α), 48kDa (β), and 30kDa (γ). Cobra venom factor (CVF) is a structural and functional analog of cobra, as well as mammalian C3. Thus, in the presence of Factor B, Factor D, and Mg2+, CVF can form a stable CVF,Bb complex that is a C3/C5 convertase enzyme; however, the CVF,Bb complex is not susceptible to regulation by Factors H and I. Four to six μg of purified CVF are equal to 1.0 unit of functional activity as measured by the method of Cochrane, et al.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Cobra Venom Factor
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species N. kaouthia
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.