Learn More
Description
- Unique pH test papers feature covalently bound indicators that will not bleed, dilute, or contaminate trace samples
- Strips give sharp, clear color changes for easy comparison to color scale provided on the colorpHast package Full-range (Universal) Strips measure 0 to 14pH range in a single test with 0.5pH sensitivity—each strip includes four squares with different indicators Intermediate-range Strips with three squares each have a sensitivity of 0.3 to 0.5 Narrow-range Strips with one pad have a sensitivity of 0.2 to 0.3
Order Info
Each pack contains 100 strips
Specifications
Specifications
| pH Range | 0 to 14 |
| Format | Strip |
| Type | pH Indicator Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.