Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C1r, Human, Activated, Two-Chain Form, Protein

Catalog No. 204878250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-487-8250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-487-8250UG Supplier MilliporeSigma™ Supplier No. 204878250UG
Add to Cart
Add to Cart
Native, human, activated, C1r complement component. Generally the non-activated form composed of a single-chain glycoprotein. Present in normal human serum at 34μg/mL. Non-activated C1r is found in circulating blood plasma as a dimer that associates with one C1q molecule and two C1s molecules to form the first component of complement (C1). Ensuing C1q binding to classical complement pathway activators, each C1r proenzyme monomer is activated by cleavage into two disulfide-linked fragments of M.W. 60,000 and 35,000. The 35kDa peptide contains C1r enzymatic active site. Activated C1r continues activation via classical pathway by cleaving, and thus activating C1s.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 140mM NaCl, 10mM imidazole, 2mM EDTA, pH 7.4.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C1r
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Structural Form Two-Chain Form
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.