Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C3, Human, Protein

Catalog No. 204885250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-5250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-5250UG Supplier MilliporeSigma™ Supplier No. 204885250UG
Add to Cart
Add to Cart
Native, human C3 complement component. Glycoprotein composed of two non-identical disulfide-bonded subunits with M.W. of 115kDa (α) and 75kDa (β). Present in normal human serum at 1.25mg/mL. Classical and alternative activation pathways of complement converge at C3 step. Activation via either pathway can result in assembly of C3-cleaving enzymes (C3 convertases) on target surfaces. Both C3 convertases cleave the C3 α-chain at peptide bond 77 resulting in production of C3a (M.W. 9083) and C3b fragments (M.W. 180,000). Released C3a peptide is one of the three complement anaphylatoxins. The nascent C3b fragment can form a covalent ester bond with target surface. This covalent attachment of C3b to target acceptors is required for continuation of complement activation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C3
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.