Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C3a des-Arg, Human, Protein

Catalog No. 20488450UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-450UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-450UG Supplier MilliporeSigma™ Supplier No. 20488450UG
Add to Cart
Add to Cart
Native, human des-Arg C3 complement component. Single chain, 76 amino acid peptide. Once the C3a peptide has been produced in human plasma or serum, it is rapidly converted to C3a des-Arg upon removal of the C-terminal arginine by endogenous serum carboxypeptidase N. This enzymatic process is considered to be a major mechanism for controlling C3a function in vivo because C3a des-Arg has <1% of the biological activity expressed by the C3a peptide. Therefore, C3a des-Arg can serve as a negative control molecule in experiments involved with the biological activities of the intact C3a anaphylatoxin peptide.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Complement C3a des-Arg
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.