Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C4, Human, Protein

Catalog No. 204886250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-488-6250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-6250UG Supplier MilliporeSigma™ Supplier No. 204886250UG
Add to Cart
Add to Cart
Native, human C4 complement component. Glycoprotein composed of three non-identical subunits of M.W. of 93kDa (α), 75kDa (β), and 32kDa (γ) linked by disulfide bonds. Present in normal human serum at 400μg/mL. On activation of complement via the classical pathway, the C1s subcomponent of the C1 complex is converted to an active serine protease that cleaves the C4 α-chain at peptide bond 77, resulting in production of C4a (M.W. 8740) and C4b fragments (M.W. 193kDa). The released C4a peptide is one of the three complement-derived anaphylatoxins. The nascent C4b fragment can form a covalent ester bond with target surfaces. This covalent attachment of C4b to target acceptors is required for continuation of activation via classical pathway.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement C4
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.