Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C5b,6 Complex, Human, Protein

Catalog No. 20490650UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-490-650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-490-650UG Supplier MilliporeSigma™ Supplier No. 20490650UG
Add to Cart
Add to Cart
Native, human C5b,6 complement component complex. Activation of complement by either pathway results in the formation of C5-cleaving enzymes (C5 convertases) on target surfaces. The C5 convertase enzymes cleave the C5 α-chain at peptide bond 74, resulting in the formation of the C5a anaphylatoxin (M.W. 11,200) and the C5b fragment (M.W. 185,000). The nascent C5b fragment can combine with C6, C7, C8, and C9 to form the lytic C5b-9 membrane attack complex on target surfaces. When C5 activation occurs in the presence of C6 only, e.g. in C7 deficient serum, a stable C5b,6 complex is formed. In a process termed Reactive Lysis, the membrane attack complex can be assembled on target cell surfaces in the absence of any additional complement proteins by incubating target cells with the C5b,6 complex and purified C7, C8, and C9.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 120mM NaCl, 10mM HEPES, pH 7.2.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Complement C5b, 6 Complex
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.