Learn More
Description
Specifications
Specifications
| Preparation Method | Reconstitute with 5mL of sterile, distilled H2O. It is recommended that the reconstituted material be filter sterilized. |
| Quantity | 5 mL |
| Immunogen | Complement |
| Storage Requirements | Following reconstitution and sterile filtration, aliquot, and freeze (−70°C). Stock solutions are stable for up to 1 year at −70°C. |
| Regulatory Status | RUO |
| Species | Guinea Pig |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.