Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement iC3b, Human, Protein

Catalog No. 204863250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-486-3250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-486-3250UG Supplier MilliporeSigma™ Supplier No. 204863250UG
Add to Cart
Add to Cart
Native, human iC3b complement component. iC3b is formed by the cleavage of C3b by Factor I in the presence of Factor H, CR1, or membrane cofactor protein. Factor I cleavage of C3b to iC3b inactivates and prevents C3b from functioning in the C3 or C5 convertase enzymes. The iC3b fragment thus produced is a glycoprotein composed of two C3α' polypeptides of M.W. 43kDa and M.W. 63kDa which are disulfide bonded to the intact C3 β-chain (M.W. 75kDa). iC3b interaction with CR3 (CD11b/CD18) receptors present on a variety of white blood cells greatly enhances phagocytosis of iC3b coated target cells or particles.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Complement iC3b
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.