Learn More
Description
Specifications
Specifications
| Preparation Method | Reconstitute with 5mL of dH2O. |
| Quantity | 5 mL |
| Immunogen | Complement |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for 24hr. at 4°C or for up to 1 month at −20°C. |
| Regulatory Status | RUO |
| Species | Rabbit |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.