Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Creatine Phosphokinase-MM Isoenzyme, Human Skeletal Muscle, Protein

Catalog No. 23840750IU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 IU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-840-750IU 50 IU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-840-750IU Supplier MilliporeSigma™ Supplier No. 23840750IU
Add to Cart
Add to Cart
Native creatine phosphokinase-MM from human skeletal muscle. Catalyzes the conversion of creatine and ATP to phosphocreatine and ADP.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 10mM Sodium Chloride, 5mM Succinate, 5mM β-Mercaptoethanol, 1mM EDTA, 50% Glycerol, pH 7.1 ± 0.1.
Quantity 50 IU
Immunogen Creatine Phosphokinase-MM Isoenzyme
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.