Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CREBα, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17600MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-600-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-600-MI Supplier MilliporeSigma™ Supplier No. 17600
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects CREBα Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Western Blotting

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction. The ChIPAb+ CREB, antibody/primer set includes the CREB, antibody, a negative control antibody (purified rabbit IgG), and qPCR primers which amplify a 64 bp region flanking the CRE of the human cFos gene. The CREB and negative control cultured supernatant are supplied in a scalable per ChIP reaction size and can be used to functionally validate the precipitation of CREB associated chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREBα
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 25 assays per set. Recommended use: ∽10μL antibody per chromatin immunoprecipitation (dependent upon biological context).
Gene Accession No. P16220
Gene Symbols CREB1, MGC9284, CREB
Host Species Rabbit
Immunogen CREB purified antibody is made against a KLH-conjugated synthetic peptide corre-sponding to amino acids 5-24 of human CREB.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004370
Test Specificity Recognizes CREB, MW ∽43kDa.
Includes This ChIPAb+ CREB -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Rat
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Culture Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.