Learn More
MilliporeSigma™ Double Cones for Hibar™/LiChroCART™ Columns
Catalog No. M1155460001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Application):
1/16 in. Capillary Tubing with Knurled Screw
Capillary Connection O.D. 0.5 mm with Knurled Nuts
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
M1155460001 Capillary Connection O.D. 0.5 mm with Knurled Nuts
M1512380001 1/16 in. Capillary Tubing with Knurled Screw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1155460001 Supplier MilliporeSigma™ Supplier No. 1155460001
Add to Cart
Add to Cart
for capillary connections
TRUSTED_SUSTAINABILITY
Specifications
Type Double Cones
For Use With (Application) Capillary Connection O.D. 0.5 mm with Knurled Nuts
Packaging Quantity 10/Pk.
For Use With Hibar Column/LiChroCART Cartridges