Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Endo-α-N-acetylgalactosaminidase, Streptococcus pneumoniae, Recombinant Protein, E. coli

Catalog No. 32471650MIU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-471-650MIU 50 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-471-650MIU Supplier MilliporeSigma™ Supplier No. 32471650MIU
Add to Cart
Add to Cart
Recombinant, Streptococcus pneumoniae Endo-α-N-acetylgalactosaminidase expressed in E. coli. Catalyzes the hydrolysis of the unsubstituted Galβ1,3GalNAc core disaccharide attached to serine or threonine residues of glycopeptides and glycoproteins to afford free oligosaccharides. For carbohydrates containing sialic acid or fucose, pretreatment with neuraminidase or fucosidase is required.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM sodium phosphate buffer, pH 7.5.
Quantity 50 mIU
Source E. coli
Immunogen Endo-α-N-acetylgalactosaminidase
Regulatory Status RUO
Species S. pneumoniae
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.