Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Endoglycosidase F1, Elizabethkingia meningosepticum, Recombinant Protein, E. coli

Catalog No. 324725700MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
700 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-472-5700MI 700 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-472-5700MI Supplier MilliporeSigma™ Supplier No. 324725700MIU
Add to Cart
Add to Cart
Recombinant, Elizabethkingia meningosepticum endoglycosidase F1 expressed in E. coli. Cleaves asparagine-linked or free oligomannose and hybrid, but not complex oligosaccharides. Core fucosylation reduces activity by 50 fold. Endo F1 will hydrolyze sulfate containing high mannose chains. It cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine. Less sensitive to protein conformation than N-Glycosidase F and therefore is more suitable for deglycosylation of native proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 20mM Tris-HCl, pH 7.5.
Quantity 700 mIU
Source E. coli
Immunogen Endoglycosidase F1
Regulatory Status RUO
Species E. meningosepticum
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.