Learn More
MilliporeSigma™ Endothelin 1, Human and Porcine, Polypeptide, Calbiochem
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Species | Human, Porcine |
| Purification Method | HPLC |
| Purity | ≥96% |
| CAS | 117399-94-7 |
| Color | White to Off-white |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 weeks at −20°C. |
| Format | Solid |
| Formulation | Supplied as an acetate salt. |
| Product Type | Endothelin 1 |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.