Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Erythropoietin, Human, Recombinant Protein, CHO Cells

Catalog No. 32987150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-987-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-987-150UG Supplier MilliporeSigma™ Supplier No. 32987150UG
Recombinant, human erythropoietin (EPO) expressed in CHO cells. EPO is a glycoprotein produced by the kidneys and is involved in red blood cell production. It has a predicted molecular mass of ∽21kDa but migrates at ∽37kDa on SDS-PAGE.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 300mM NaCl, 20mM Na/K phosphate, pH 7.5.
Preparation Method Reconstitute with 500μL H2O or aqueous buffer.
Purification Method HPLC
Quantity 50 μg
Immunogen Erythropoietin
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C or −70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C, up to 3 months at −20°C, or up to 6 months at −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Cell Type CHO Cells
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.