Learn More
Description
Specifications
Specifications
| Content And Storage | ESGRO Supplement: (Mfr. No. CS210511): Mouse recombinant Leukemia Inhibitory Factor (LIF), 0.5 x 106U/mL. Ten (10) 0.5mL vials. MEK/GS3 Inhibitor Supplement: (Mfr. No. CS210510-500UL): Ten (10) 0.5mL. Store at -20°C up to 2 years |
| For Use With (Application) | Stem cell culture |
| Quantity | 1 Kit/5 L medium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.