Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ESGRO Complete™ Accutase™ Cell Detachment Solution

Catalog No. SF006 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF006 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF006 Supplier MilliporeSigma™ Supplier No. SF006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a cell detachment solution of proteolytic and collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.

ESGRO Complete™ Accutase™ is a cell detachment solution of proteolytic and collagenolytic enzymes that has been qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete™ Clonal Grade Medium (Cat. No. SF001-500). Accutase™ does not contain mammalian or bacterial derived products.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stable when stored at -20° C
For Use With (Application) Stem Cell Culture
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.