Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 5mM HEPES, pH 8.0. |
| Preparation Method | Reconstitute with 200μL of sterile distilled H2O |
| Purification Method | Single band by SDS-PAGE |
| Quantity | 50 μg |
| Source | Clostridium botulinum |
| Immunogen | Exoenzyme C3 |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short term storage. Upon thawing, do not re-freeze. Aqueous stock solutions are stable for up ro 2 months at 4°C or for up to 4 months at −20°C. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.