Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Express™ PLUS Membrane Filters
GSA_VA

Catalog No. GPWP04700 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Diameter (Metric):
13 mm
25 mm
47 mm
90 mm
142 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric)
GPWP04700 0.22 μm 47 mm
GPWP01300 0.22 μm 13 mm
GPWP02500 0.22 μm 25 mm
GPWP09050 0.22 μm 90 mm
GPWP14250 0.22 μm 142 mm
HPWP14250 0.45 μm 142 mm
HPWP09050 0.45 μm 90 mm
HPWP02500 0.45 μm 25 mm
HPWP04700 0.45 μm 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. GPWP04700 Supplier MilliporeSigma™ Supplier No. GPWP04700
Add to Cart
Add to Cart

Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features and Benefit:
  • Ultra fast filtration and high throughput shorten process time
  • Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
  • Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color White
Disposable Reusable
Filter Type Screen Filter
Format Disc
Material PES
Pore Size 0.22 μm
Sterility Non-sterile
Flow Rate 15 (at 690 Mbar (10 psi))
For Use With (Application) Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Hydrophobicity Hydrophilic
Membrane Type Plain
Type Filter Membranes
Wettability Hydrophilic
Diameter (English) 1.9 in.
Diameter (Metric) 47 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.