Learn More
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
Features and Benefit:- Ultra fast filtration and high throughput shorten process time
- Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
- Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | White |
| Disposable | Reusable |
| Filter Type | Screen Filter |
| Format | Disc |
| Material | PES |
| Pore Size | 0.45 μm |
| Sterility | Non-sterile |
| Flow Rate | 23 (at 690 Mbar (10 psi)) |
| For Use With (Application) | Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
