Learn More
MilliporeSigma™ EXtrelut NT Prepacked Column: Cannulae
Catalog No. M1153730001 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging Quantity:
100 Units
Catalog No. Packaging Quantity
M1153730001 100 Units
1 options
Catalog No. M1153730001 Supplier MilliporeSigma™ Supplier No. 1.15373.0001
for EXtrelut™ NT 1 and EXtrelut™ NT 3
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
TRUSTED_SUSTAINABILITY
Specifications
Type Cannula
Packaging Quantity 100 Units
For Use With EXtrelut NT1 and NT3 Prepacked Columns