Learn More
MilliporeSigma™ EXtrelut™ NT Prepacked Columns
pre-packed columns for extraction of lipophilic compounds from aqueous solutions
$446.00 - $790.00
Specifications
| For Use With (Application) | To Obtain High Degree Purity |
|---|---|
| Sorbent | SPE Sorbent |
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|
M1150940001
|
MilliporeSigma™
1150940001 |
EXtrelut NT 1 Prepacked Column, Glass |
Each for $534.00
|
|
|||||
|
M1150950001
|
MilliporeSigma™
1.15095.0001 |
EXtrelut NT 3 Prepacked Column, Glass |
Each for $446.00
|
|
|||||
|
M1150960001
|
MilliporeSigma™
1150960001 |
EXtrelut NT 20 Prepacked Column, Polyethylene |
Each for $790.00
|
|
|||||
Description
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.Specifications
| To Obtain High Degree Purity | |
| SPE Sorbent |