Learn More
MilliporeSigma™ EZ-Fit™ Manifolds
Catalog No. EZFITH0LD6 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Microfil V or Microfil S filtration units
Microfil funnels and membranes
Rubber Stopper Tulip
Manifold Type:
1-place
3-places
6-places
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Manifold Type
EZFITH0LD6 Rubber Stopper Tulip 6-places
EZFITMIC01 Microfil funnels and membranes 1-place
EZFITH0LD1 Rubber Stopper Tulip 1-place
EZFITH0LD3 Rubber Stopper Tulip 3-places
EZFITL0W03 Microfil V or Microfil S filtration units 3-places
EZFITMIC03 Microfil funnels and membranes 3-places
EZFITL0W06 Microfil V or Microfil S filtration units 6-places
EZFITMIC06 Microfil funnels and membranes 6-places
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. EZFITH0LD6 Supplier MilliporeSigma™ Supplier No. EZFITH0LD6
Add to Cart
Add to Cart
Request a Quote
General microbiological analysis
EZ-Fit™ Manifold is available with the new Microfil™ head for use with presterilized, ready-to-use funnels and membranes.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With (Equipment) Rubber Stopper Tulip
Material Stainless Steel
Manifold Type 6-places