Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Factor B, Human, Protein

Catalog No. 341262250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-126-2250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-126-2250UG Supplier MilliporeSigma™ Supplier No. 341262250UG
Add to Cart
Add to Cart
Native factor B from human serum. Single-chain glycoprotein present in normal human serum at about 200μg/mL. During activation of complement via the alternative pathway, Factor B is cleaved and thus activated by Factor D into two fragments of M.W. of 30kDa (Ba) and 63kDa (Bb). The Bb fragment contains the serine protease enzymatic active site. The activated Bb fragment continues activation via the alternative pathway by cleaving, and thus activating, C3 and C5.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In PBS, pH 7.2.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen Factor B
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.