Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ferritin, Human Liver, Protein

Catalog No. 3414821MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-148-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-148-21MG Supplier MilliporeSigma™ Supplier No. 3414821MG
Add to Cart
Add to Cart
Native ferritin from human liver. Sterile-filtered. A major iron storage protein. Suitable for use in immunoassays, as an immunogen, and in enzyme/radiolabeling.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 0.9% NaCl, 0.1% NaN3, pH 7.0.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Ferritin
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.