Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibrinogen, Bovine Plasma, Protein

Catalog No. 3415731GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-157-31GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-157-31GM Supplier MilliporeSigma™ Supplier No. 3415731GM
Add to Cart
Add to Cart
Native fibrinogen from bovine plasma. An acute phase plasma glycoprotein synthesized and secreted by hepatic parenchymal cells. Contains ≥95% clottable proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 20mM sodium citrate-HCl, pH 7.4.
Preparation Method Sterile dH2O
Purification Method Homogeneous by SDS-PAGE
Quantity 1 g
Immunogen Fibrinogen
Storage Requirements Following reconstitution, aliquot and freeze (−20°C) for long term storage or store at room temperature (20°C) for short term storage. Do not store solutions on ice; precipitation will occur. Stock solutions are stable for up to 4 h at 20°C or for up to 6 months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.