Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibrinogen, Fragment D, Human Plasma, Protein

Catalog No. 341600200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-160-0200UG 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-160-0200UG Supplier MilliporeSigma™ Supplier No. 341600200UG
Add to Cart
Add to Cart
Native fibrinogen fragment D from human plasma prepared by plasmin digestion of purified fibrinogen followed by preparative electrophoresis. Thermolabile core fragment of fibrinogen. Complete proteolysis of a single fibrinogen molecule produces two D and one E fragments. The D and E fragments have no common antigenic determinants; therefore, no cross-reaction occurs. Reported to increase endothelial membrane permeability to albumin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 400mM glycine, 150mM NaCl.
Preparation Method Sterile distilled H2O (200μg/mL)
Purification Method Single band by SDS-PAGE
Quantity 200 μg
Immunogen Fibrinogen, Fragment D
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 4hr. at 20°C or for up to 8hr. at 4°C or for up to 3 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.