Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibroblast Growth Factor-Basic Human Recombinant Protein
GSA_VA

Catalog No. GF003 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF003 50 μg
GF003AFMG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. GF003 Supplier MilliporeSigma™ Supplier No. GF003
Add to Cart
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture, Stem Cell Culture
Name Fibroblast Growth Factor-basic (FGF-b)
Quantity 50 μg
Source Expressed in E. coli
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade Greater than 95% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.