Learn More
MilliporeSigma™ Filter Holder Vacuum Manifold
GSA_VA
Catalog No. XX2504700 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Positions:
3
6
Material:
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Positions Material
XX2504700 6 Stainless Steel
XX2504735 3 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. XX2504700 Supplier MilliporeSigma™ Supplier No. XX2504700
Filter Holder Vacuum Manifold provides vacuum support for simultaneous filtration of either three or six test samples.
  • Use Millipore glass, plastic or stainless-steel filter holders fitted with standard No. 8 perforated stoppers over the holder outlets.
  • Manifolds connect to single vacuum source and use separate control valves at each station for independent operation.
  • Use vent ports on each stainless-steel manifold station to attach Swinnex™ filter holders or Millex™ filter units to maintain sterility on the test filter's downstream side.
Stainless-steel Manifolds:
  • Stainless-steel body, three-way valves, filter holder supports, anodized aluminum handles, stainless-steel hose connector, pipe plug.
  • Units are autoclavable at 121°C (250°F).
PVC Manifold:
  • Polyvinylchloride body, stainless-steel two-way valve, polypropylene hose adapters and handles.
  • Not autoclavable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Manifold
For Use With (Equipment) Vacuum Filter Holder
Material Stainless Steel
Sterility Non-sterile
No. of Positions 6
Type Manifold