Learn More
MilliporeSigma™ Flow-limiting Orifices
Catalog No. XX5000020 Shop All MilliporeSigma Products
Change view
Click to view available options
Airflow Rate:
1 L/min.
2 L/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Airflow Rate
XX5000020 2 L/min.
XX5000002 1 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. XX5000020 Supplier MilliporeSigma™ Supplier No. XX5000020
  • $202.00
    $177.65 / Each of 1
    Save $24.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Creates quantitative airflow through a filter for airborne contamination analysis
Flow Limiting Orifices create quantitative airflow through a MF-Millipore filter for airborne contamination analysis. The orifices are threaded into the outlet of a Millipore Aerosol Analysis Holder or Aerosol Adapter to enable a constant flow rate when required vacuum level is applied.Ratings are accurate to within approximately 15%. For greater precision, calibrate orifice with a flowmeter at actual working vacuum level.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Environmental Test Kits or Supplies
Material Stainless Steel
For Use With (Equipment) Millipore Aerosol Analysis Holder or Aerosol Adapter
For Use With (Application) To create quantitative airflow through a Millipore filter for airborne contamination analysis
Airflow Rate 2 L/min.
Includes Flow-limiting Orifice, 2 L/min