Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Follicle Stimulating Hormone, Human Pituitary, Iodination Grade, Protein

Catalog No. 86900110UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-900-110UG 10 μg
86-900-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-900-110UG Supplier MilliporeSigma™ Supplier No. 86900110UG
Add to Cart
Add to Cart
Native, human follicle stimulating hormone from pituitary gland. Two-chain glycoprotein gonadotropic hormone that induces maturation of the Graafian follicles of the ovary. Promotes the development of germinal cells in males. FSH regulates Sertoli cells by acting on G-protein-linked cell surface FSH receptors. Activates cytosolic soluble protein tyrosine kinase (CyPTk). Immunopotency: ≥2000IU/mg (WHO 1st IRP 69/104).
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method PBS
Quantity 10 μg
Immunogen Follicle Stimulating Hormone
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥90%, Iodination Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.