Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FPTase, Rat, Recombinant Protein, E. coli

Catalog No. 344145100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-414-5100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-414-5100UG Supplier MilliporeSigma™ Supplier No. 344145100UG
Add to Cart
Add to Cart
Recombinant, rat FPTase expressed in E. coli. A heterodimeric enzyme that catalyzes the transfer of a 15-carbon isoprenoid group to a variety of cellular proteins including Ras. Recognizes the C-terminal CAAX cysteine motif where the farnesylated cysteine is followed by two aliphatic amino acids and either leucine or serine.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 40 mM NaCl, 25mM HEPES, 5mM DTT, pH 7.2.
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Immunogen FPTase
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Rat
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.