Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| For Use With (Application) | Immunoblotting |
| Formulation | In 50mM HEPES, 1mM DTT, 1mM EDTA, 0.1% LUBROL™ Detergent, pH 7.6. |
| Quantity | 50μg |
| Immunogen | G-Proteins |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Bovine |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.