Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GDF-5, Mouse, Recombinant Protein

Catalog No. 34573150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-573-150UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-573-150UG Supplier MilliporeSigma™ Supplier No. 34573150UG
Add to Cart
Add to Cart
Recombinant, mouse growth differentiation factor 5 (GDF-5) consisting of amino acids 376-495, expressed in E. coli with and without an N-terminal methionine. GDF-5, a member of the bone morphogenetic protein family, is involved in regulation of myogenesis, bone morphogenesis, neuron differentiation and survival, and chondrogenesis regulation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM filter-sterilized Acetonitrile, TFA, BSA.
Preparation Method Reconstitute to 150μg/mL in sterile 4mM HCl containing at least 0.1% human or bovine serum albumin.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen GDF-5
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Solid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.