Learn More
Description
Specifications
Specifications
| Species | Mouse |
| Purification Method | SDS-PAGE |
| Purity | ≥90% |
| Content And Storage | Following reconstituyion refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C. |
| Format | Lyophilized |
| Formulation | Lyophilized from 0.2μM filtered PBS, BSA. |
| Peptide Tag | His·Tag™ |
| Product Type | GDF-8 Propeptide |
| Quantity | 25 μg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.